Ethyl undecafluoroamyl ketone ≥98.0%

Supplier: TCI America

Synonyms: 1H,1H,1H,2H,2H-Undecafluoro-3-octanoneEthyl Perfluoroamyl KetoneEthyl Undecafluoropentyl Ketone4,4,5,5,6,6,7,7,8,8,8-Undecafluoro-3-octanone

P1363-5G
TCP1363-005GEA 250 USD
TCP1363-005G
Ethyl undecafluoroamyl ketone ≥98.0%
Ethyl undecafluoroamyl ketone

CAS Number: 383177-55-7
MDL Number: MFCD02093488
Molecular Formula: C8H5F11O
Molecular Weight: 326.11
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.53


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds
Other Product Names / Synonyms: 1H,1H,1H,2H,2H-Undecafluoro-3-octanone, Ethyl Perfluoroamyl Ketone, Ethyl Undecafluoropentyl Ketone, 4,4,5,5,6,6,7,7,8,8,8-Undecafluoro-3-octanone

Formula: C₈H₅F₁₁O
MW: 326.11 g/mol
Density: 1.53 (20 °C)
MDL Number: MFCD02093488
CAS Number: 383177-55-7

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR