ɑ-Pinene oxide ≥95.0%

Supplier: TCI America

Synonyms: 2,3-Epoxypinane

P1362-25ML P1362-250ML
TCP1362-250MLEA 218.13 USD
TCP1362-250ML TCP1362-025ML
ɑ-Pinene oxide ≥95.0%
alpha-Pinene oxide

CAS Number: 1686-14-2
MDL Number: MFCD00066955
Molecular Formula: C10H16O
Molecular Weight: 152.24
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 66
Specific Gravity (20/20): 0.97
Specific rotation [a]20/D: -55 deg (neat)


Product Categories: Bioscience, Bicyclic Monoterpenes
Other Product Names / Synonyms: 2,3-Epoxypinane

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Density: 0.97 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00066955
CAS Number: 1686-14-2
https://us.vwr.com/stibo/web/std.lang.all/20/10/28992010.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR