Diethyl-1,4-phenylenediacrylate

Supplier: TCI America

Synonyms: 1,4-Phenylenediacrylic Acid Diethyl Ester

P1345-5G
TCP1345-005GEA 473 USD
TCP1345-005G
Diethyl-1,4-phenylenediacrylate
Diethyl-1,4-phenylenediacrylate

CAS Number: 17088-28-7
MDL Number: MFCD00017521
Molecular Formula: C16H18O4
Molecular Weight: 274.32
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 95


Other Product Names / Synonyms: 1,4-Phenylenediacrylic Acid Diethyl Ester

Formula: C₁₆H₁₈O₄
MW: 274.32 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00017521
CAS Number: 17088-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR