Monohexyl phthalate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Phthalic Acid Monohexyl Ester

P1313-5G
TCP1313-005GEA 152.19 USD
TCP1313-005G
Monohexyl phthalate ≥98.0% (by GC, titration analysis)
Monohexyl phthalate

CAS Number: 24539-57-9
MDL Number: MFCD00517703
Molecular Formula: C14H18O4
Molecular Weight: 250.29
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Freezing point (°C): 12
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Phthalic Acid Monohexyl Ester

Formula: C₁₄H₁₈O₄
MW: 250.29 g/mol
MDL Number: MFCD00517703
CAS Number: 24539-57-9
https://us.vwr.com/stibo/web/std.lang.all/19/71/28991971.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR