Allyltris(trimethylsilyloxy)silane ≥96.0%

Supplier: TCI America

Synonyms: 3-Allyl-1,1,1,5,5,5-hexamethyl-3-((trimethylsilyl)oxy)trisiloxane

A2111-1G A2111-5G
TCA2111-5GEA 402 USD
TCA2111-5G TCA2111-1G
Allyltris(trimethylsilyloxy)silane ≥96.0%
Allyltris(trimethylsilyloxy)silane

CAS Number: 7087-21-0
MDL Number: MFCD00054804
Molecular Formula: C12H32O3Si4
Molecular Weight: 336.73
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 111
Flash Point (°C): 67
Specific Gravity (20/20): 0.87

Formula: C₁₂H₃₂O₃Si₄
MW: 336.73 g/mol
Boiling Pt: 111 °C
Density: 0.87 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00054804
CAS Number: 7087-21-0
https://us.vwr.com/stibo/web/std.lang.all/02/75/28930275.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR