Ethyl-4-phenoxybutyrate ≥95.0%

Supplier: TCI America

Synonyms: 4-Phenoxybutyric Acid Ethyl Ester

P1286-25G
TCP1286-025GEA 229 USD
TCP1286-025G
Ethyl-4-phenoxybutyrate ≥95.0%
Ethyl-4-phenoxybutyrate

CAS Number: 2364-59-2
MDL Number: MFCD01321182
Molecular Formula: C12H16O3
Molecular Weight: 208.26
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 173
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: 4-Phenoxybutyric Acid Ethyl Ester

Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
Boiling Pt: 173 °C
Density: 1.06 (20 °C)
MDL Number: MFCD01321182
CAS Number: 2364-59-2
https://us.vwr.com/stibo/web/std.lang.all/19/17/28991917.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR