Diethyl-1-pyrrolidinemethylphosphonate ≥95.0%

Supplier: TCI America

Synonyms: 1-Pyrrolidinemethylphosphonic Acid Diethyl Ester

P1258-5G
TCP1258-005GEA 112.62 USD
TCP1258-005G
Diethyl-1-pyrrolidinemethylphosphonate ≥95.0%
Diethyl-1-pyrrolidinemethylphosphonate

CAS Number: 51868-96-3
MDL Number: MFCD00040516
Molecular Formula: C9H20NO3P
Molecular Weight: 221.24
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 103
Specific Gravity (20/20): 1.06


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: 1-Pyrrolidinemethylphosphonic Acid Diethyl Ester

Formula: C₉H₂₀NO₃P
MW: 221.24 g/mol
Density: 1.06 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD00040516
CAS Number: 51868-96-3
https://us.vwr.com/stibo/web/std.lang.all/18/91/28991891.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR