9-Phenyl-9-fluorenol ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
9-Phenyl-9H-fluoren-9-ol
, 9-Hydroxy-9-phenylfluorene
P1231-5G
TCP1231-005GEA
101.45
USD
TCP1231-005G
9-Phenyl-9-fluorenol ≥98.0% (by HPLC)
9-Phenyl-9-fluorenol
CAS Number: 25603-67-2
MDL Number: MFCD00037121
Molecular Formula: C19H14O
Molecular Weight: 258.32
Purity/Analysis Method:
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 111
Other Product Names / Synonyms: 9-Hydroxy-9-phenylfluorene
Formula:
C₁₉H₁₄O MW: 258.32 g/mol Melting Pt: 111 °C |
MDL Number:
MFCD00037121 CAS Number: 25603-67-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...