Monobenzyl phthalate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Phthalic Acid Monobenzyl EsterBenzyl Hydrogen Phthalate2-((Benzyloxy)carbonyl)benzoic acid

P1191-10G
TCP1191-010GEA 212 USD
TCP1191-010G
Monobenzyl phthalate ≥98.0% (by GC, titration analysis)
Monobenzyl phthalate

CAS Number: 2528-16-7
MDL Number: MFCD00020279
Molecular Formula: C15H12O4
Molecular Weight: 256.26
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 106


Other Product Names / Synonyms: Benzyl Hydrogen Phthalate, Phthalic Acid Monobenzyl Ester

Formula: C₁₅H₁₂O₄
MW: 256.26 g/mol
Melting Pt: 106 °C
MDL Number: MFCD00020279
CAS Number: 2528-16-7
https://us.vwr.com/stibo/web/std.lang.all/18/24/28991824.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR