Diethyl phthalimidomalonate ≥96.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Phthalimidomalonic Acid Diethyl Ester

P1180-5G
TCP1180-005GEA 99.47 USD
TCP1180-005G
Diethyl phthalimidomalonate ≥96.0% (by total nitrogen basis)
Diethyl phthalimidomalonate

CAS Number: 5680-61-5
MDL Number: MFCD00065025
Molecular Formula: C15H15NO6
Molecular Weight: 305.29
Purity/Analysis Method: 96.0% (N)
Form: Crystal
Color: White
Melting point (°C): 76


Other Product Names / Synonyms: Phthalimidomalonic Acid Diethyl Ester

Formula: C₁₅H₁₅NO₆
MW: 305.29 g/mol
Melting Pt: 76 °C
MDL Number: MFCD00065025
CAS Number: 5680-61-5
https://us.vwr.com/stibo/web/std.lang.all/18/09/28991809.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR