2-Propyl-2-imidazoline

Supplier: TCI America

P1164-25G
TCP1164-025GEA 160.64 USD
TCP1164-025G
2-Propyl-2-imidazoline
2-Propyl-2-imidazoline

CAS Number: 15450-05-2
MDL Number: MFCD00191487
Molecular Formula: C6H12N2
Molecular Weight: 112.18
Form: Crystal
Color: Pale Yellow
Boiling point (°C): 140
Melting point (°C): 42

Formula: C₆H₁₂N₂
Boiling Pt: 140 °C
Melting Pt: 42 °C
MDL Number: MFCD00191487
CAS Number: 15450-05-2
https://us.vwr.com/stibo/web/std.lang.all/17/91/28991791.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR