5-Phenyl-4-pentyn-1-ol ≥98.0%

Supplier: TCI America

Synonyms: 5-Phenylpent-4-yn-1-ol

P1158-1G
TCP1158-001GEA 285.12 USD
TCP1158-001G
5-Phenyl-4-pentyn-1-ol ≥98.0%
5-Phenyl-4-pentyn-1-ol

CAS Number: 24595-58-2
MDL Number: MFCD00191481
Molecular Formula: C11H12O
Molecular Weight: 160.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow

Formula: C₁₁H₁₂O
MW: 160.22 g/mol
MDL Number: MFCD00191481
CAS Number: 24595-58-2
https://us.vwr.com/stibo/web/std.lang.all/17/86/28991786.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR