(Perfluoro-n-octyl)phenyliodonium trifluoromethanesulfonate
Synonyms:
(Eicosafluoro-n-octyl)phenyliodonium Trifluoromethanesulfonate
P1081-1G
TCP1081-001GEA
415.09
USD
TCP1081-001G
(Perfluoro-n-octyl)phenyliodonium trifluoromethanesulfonate
(Perfluoro-n-octyl)phenyliodonium trifluoromethanesulfonate
CAS Number: 77758-89-5
MDL Number: MFCD00135120
Molecular Formula: C15H5F20IO3S
Molecular Weight: 772.13
Form: Crystal
Color: White
Storage Temperature: 0-10°C
Other Product Names / Synonyms: (Eicosafluoro-n-octyl)phenyliodonium Trifluoromethanesulfonate
Formula:
C₁₅H₅F₂₀IO₃S MW: 772.13 g/mol |
MDL Number:
MFCD00135120 CAS Number: 77758-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...