(Perfluorohexyl)phenyliodonium trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)

Synonyms: (Tridecafluorohexyl)phenyliodonium Trifluoromethanesulfonate

P1080-5G P1080-1G
TCP1080-5GEA 1054.72 USD
TCP1080-5G TCP1080-001G
(Perfluorohexyl)phenyliodonium trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
(Perfluorohexyl)phenyliodonium trifluoromethanesulfonate

CAS Number: 77758-84-0
MDL Number: MFCD00135119
Molecular Formula: C13H5F16IO3S
Molecular Weight: 672.12
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Hypervalent Iodine Compounds, Fluorous Compounds, Perfluoroalkylating Agents
Other Product Names / Synonyms: (Tridecafluorohexyl)phenyliodonium Trifluoromethanesulfonate

Formula: C₁₃H₅F₁₆IO₃S
MW: 672.12 g/mol
MDL Number: MFCD00135119
CAS Number: 77758-84-0
https://us.vwr.com/stibo/web/std.lang.all/17/11/28991711.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR