1-Iodo-1-propyne ≥97.0%

Supplier: TCI America

Synonyms: 1-Propyn-1-yl Iodide

P1065-1G
TCP1065-001GEA 546.84 USD
TCP1065-001G
1-Iodo-1-propyne ≥97.0%
1-Iodo-1-propyne

CAS Number: 624-66-8
MDL Number: MFCD00059101
Molecular Formula: C3H3I
Molecular Weight: 165.96
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 110
Specific Gravity (20/20): 1.99
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Langmuir-Blodgett(LB) Film Forming Agents
Other Product Names / Synonyms: 1-Propyn-1-yl Iodide

Formula: C₃H₃I
MW: 165.96 g/mol
Boiling Pt: 110 °C
Density: 1.99 (20 °C)
MDL Number: MFCD00059101
CAS Number: 624-66-8
https://us.vwr.com/stibo/web/std.lang.all/16/99/28991699.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR