2-Phenyl-1,3-indandione ≥98.0%

Synonyms: 2-Phenyl-1H-indene-1,3(2H)-dionePhenindione

P1029-250G
TCP1029-250GEA 386.58 USD
TCP1029-250G
2-Phenyl-1,3-indandione ≥98.0%
2-Phenyl-1,3-indandione

CAS Number: 83-12-5
MDL Number: MFCD00003782
Molecular Formula: C15H10O2
Molecular Weight: 222.24
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 148


Other Product Names / Synonyms: Phenindione

Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Melting Pt: 148 °C
MDL Number: MFCD00003782
CAS Number: 83-12-5
https://us.vwr.com/stibo/web/std.lang.all/16/65/28991665.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR