TCIAP1003

P1003-1G
TCP1003-001GEA 116.52 USD
TCP1003-001G
TCIAP1003
CAS Number: 1074-29-9
MDL Number: MFCD00000460
Molecular Formula: C6H5Cl3Ge
Molecular Weight: 256.09
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 106
Flash Point (°C): 110
Specific Gravity (20/20): 1.58

Other Product Names / Synonyms: Phenyltrichlorogermane
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR