Disodium phthalocyanine
Supplier: TCI America
Synonyms:
Phthalocyanine Sodium
P0973-1G
P0973-5G
TCP0973-001GEA
72.47
USD
TCP0973-001G
TCP0973-005G
Disodium phthalocyanine
Disodium phthalocyanine
CAS Number: 25476-27-1
MDL Number: MFCD00066368
Molecular Formula: C32H16N8Na2
Molecular Weight: 558.52
Form: Crystal
Color: Deep Blue
Other Product Names / Synonyms: Phthalocyanine Sodium, Sodium Phthalocyanine, Phthalocyanine Disodium
Formula:
C₃₂H₁₆N₈Na₂ MW: 558.52 g/mol |
MDL Number:
MFCD00066368 CAS Number: 25476-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...