cis-3-Hexenyl pyruvate ≥95.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Pyruvic Acid cis-3-Hexenyl Ester

P0894-25ML
TCP0894-025MLEA 120.74 USD
TCP0894-025ML
cis-3-Hexenyl pyruvate ≥95.0% (by GC, titration analysis)
cis-3-Hexenyl pyruvate

CAS Number: 68133-76-6
MDL Number: MFCD00036527
Molecular Formula: C9H14O3
Molecular Weight: 170.21
Purity/Analysis Method: 95.0% (GC,T)
Form: Clear Liquid
Color: Slightly Pale Yellow
Boiling point (°C): 98
Flash Point (°C): 108
Specific Gravity (20/20): 0.99
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Pyruvic Acid cis-3-Hexenyl Ester

Formula: C₉H₁₄O₃
MW: 170.21 g/mol
Boiling Pt: 98 °C
Density: 0.99 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00036527
CAS Number: 68133-76-6
https://us.vwr.com/stibo/web/std.lang.all/15/66/28991566.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR