Risocaine (propyl-4-aminobenzoate) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Propyl 4-aminobenzoate
A0274-25G
TCA0274-025GEA
63
USD
TCA0274-025G
Risocaine (propyl-4-aminobenzoate) ≥98.0% (by titrimetric analysis)
Risocaine (Propyl 4-aminobenzoate)
CAS Number: 94-12-2
MDL Number: MFCD00017111
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 74
Other Product Names / Synonyms: 4-Aminobenzoic Acid Propyl Ester
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...