2-Propylvaleryl chloride ≥99.0%
Supplier: TCI America
Synonyms:
2-Propylpentanoyl chloride
, 2,2-Di-n-propylacetylchloride
P0824-5ML
TCP0824-005MLEA
169
USD
TCP0824-005ML
2-Propylvaleryl chloride ≥99.0%
2-Propylvaleryl chloride
CAS Number: 2936-08-5
MDL Number: MFCD00051446
Molecular Formula: C8H15ClO
Molecular Weight: 162.66
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 132
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: Dipropylacetyl Chloride, 2-Propylvaleroyl Chloride, Valproyl Chloride
Formula:
C₈H₁₅ClO MW: 162.66 g/mol Boiling Pt: 132 °C Density: 0.95 (20 °C) |
MDL Number:
MFCD00051446 CAS Number: 2936-08-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...