(R)-1-Acetyl-3-pyrrolidinol ≥98.0%

Supplier: TCI America

Synonyms: (R)-1-Acetyl-3-hydroxypyrrolidine

A2049-1G
TCA2049-1GEA 389 USD
TCA2049-1G
(R)-1-Acetyl-3-pyrrolidinol ≥98.0%
(R)-1-Acetyl-3-pyrrolidinol

CAS Number: 916733-17-0
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 83
Specific rotation [a]20/D: -42 deg (C=3, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-1-Acetyl-3-hydroxypyrrolidine

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: 83 °C
MDL Number: MFCD09909313
CAS Number: 916733-17-0
https://us.vwr.com/stibo/web/std.lang.all/01/93/28930193.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR