Polyethylene glycol mono-4-nonylphenyl ether, M.W. n=∼2
Supplier: TCI America
P0704-25G
TCP0704-25GEA
34.51
USD
TCP0704-25G
Polyethylene glycol mono-4-nonylphenyl ether, M.W. n=∼2
Polyethylene glycol mono-4-nonylphenyl ether
CAS Number: 26027-38-3
MDL Number: MFCD00148006
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 100
Specific Gravity (20/20): 1.00
Product Categories: Materials Chemistry, Polymers, Nonionic Surfactants
Density:
1.00 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00148006 CAS Number: 26027-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...