Bis(trans-3,3,5-trimethylcyclohexyl)phthalate ≥95.0%

Supplier: TCI America

Synonyms: Phthalic Acid Bis(trans-3,3,5-trimethylcyclohexyl) Ester

P0697-25G
TCP0697-025GEA 956 USD
TCP0697-025G
Bis(trans-3,3,5-trimethylcyclohexyl)phthalate ≥95.0%
Bis(trans-3,3,5-trimethylcyclohexyl)phthalate

CAS Number: 245652-82-8
MDL Number: MFCD00070481
Molecular Formula: C26H38O4
Molecular Weight: 414.59
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 54


Other Product Names / Synonyms: Phthalic Acid Bis(trans-3,3,5-trimethylcyclohexyl) Ester

Formula: C₂₆H₃₈O₄
MW: 414.59 g/mol
Melting Pt: 54 °C
MDL Number: MFCD00070481
CAS Number: 245652-82-8
https://us.vwr.com/stibo/web/std.lang.all/14/01/28991401.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR