cis-1,3-Pentadiene ≥97.0% stabilized

Supplier: TCI America

Synonyms: cis-Piperylene

P0650-5G P0650-1G
TCP0650-1GEA 300.35 USD
TCP0650-1G TCP0650-005G
cis-1,3-Pentadiene ≥97.0% stabilized
cis-1,3-Pentadiene

(stabilized with TBC)
CAS Number: 1574-41-0
MDL Number: MFCD00063977
Molecular Formula: C5H8
Molecular Weight: 68.12
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 43
Melting point (°C): -140
Flash Point (°C): -28
Specific Gravity (20/20): 0.69
Storage Temperature: 0°C


Other Product Names / Synonyms: cis-Piperylene

Formula: C₅H₈
MW: 68.12 g/mol
Boiling Pt: 43 °C
Melting Pt: –140 °C
Density: 0.69 (20 °C)
Flash Pt: –28 °C
MDL Number: MFCD00063977
CAS Number: 1574-41-0
EINECS: 216-401-0
UN: 3295
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/13/69/28991369.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR