Ponceau Xylidine
Supplier: TCI America
Synonyms:
Xylidine ponceau
P0590-25G
TCP0590-025GEA
70.18
USD
TCP0590-025G
Ponceau Xylidine
Ponceau Xylidine
CAS Number: 3761-53-3
MDL Number: MFCD00003897
Molecular Formula: C18H16N2O7S2
Molecular Weight: 480.42
Form: Crystal
Color: Red
Other Product Names / Synonyms: Ponceau de Xylidine, Ponceau R
Formula:
C₁₈H₁₆N₂O₇S₂ MW: 480.42 g/mol |
MDL Number:
MFCD00003897 CAS Number: 3761-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...