1-(4-Nitrophenylazo)-2-naphthol

Supplier: TCI America

Synonyms: Para Red1-((4-Nitrophenyl)diazenyl)naphthalen-2-ol

P0586-25G
TCP0586-025GEA 45.68 USD
TCP0586-025G
1-(4-Nitrophenylazo)-2-naphthol
1-(4-Nitrophenylazo)-2-naphthol

CAS Number: 6410-10-2
MDL Number: MFCD00003908
Molecular Formula: C16H11N3O3
Molecular Weight: 293.28
Form: Crystal
Color: Yellow Red
Melting point (°C): 252
Lambda max.: 487 nm (Toluene)


Other Product Names / Synonyms: 1-(4-Nitrophenylazo)-2-naphthol

Formula: C₁₆H₁₁N₃O₃
MW: 293.28 g/mol
Melting Pt: 252 °C
MDL Number: MFCD00003908
CAS Number: 6410-10-2
https://us.vwr.com/stibo/web/std.lang.all/13/19/28991319.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR