1-Amino-1-cyclobutanecarboxylic acid hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A2032-5G
TCA2032-5GEA 152 USD
TCA2032-5G
1-Amino-1-cyclobutanecarboxylic acid hydrochloride ≥98.0% (by titrimetric analysis)
1-Amino-1-cyclobutanecarboxylic acid hydrochloride

CAS Number: 98071-16-0
Molecular Formula: C5H9NO2
Molecular Weight: 151.59
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 226

Formula: C₅H₁₀ClNO₂
MW: 151.5914 g/mol
Melting Pt: 226 °C
MDL Number: MFCD00661068
CAS Number: 98071-16-0
https://us.vwr.com/stibo/web/std.lang.all/01/79/28930179.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR