Pyridine-2-carbohydrazide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Picolinic acid hydrazide2-Pyridinecarboxylic acid hydrazide2-Picolinyl hydrazide2-PyridinecarbohydrazidePicolinohydrazide

P0426-25G
TCP0426-025GEA 264.92 USD
TCP0426-025G
Pyridine-2-carbohydrazide ≥98.0% (by titrimetric analysis)
Pyridine-2-carbohydrazide

CAS Number: 1452-63-7
MDL Number: MFCD00059782
Molecular Formula: C6H7N3O
Molecular Weight: 137.14
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 100


Other Product Names / Synonyms: 2-Picolinyl Hydrazide, Picolinic Acid Hydrazide

Formula: C₆H₇N₃O
MW: 137.1393 g/mol
Melting Pt: 100 °C
MDL Number: MFCD00059782
CAS Number: 1452-63-7
https://us.vwr.com/stibo/web/std.lang.all/11/20/28991120.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR