(R)-(−)-Phenylephrine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(R)-(-)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol hydrochloride
CAS Number: 61-76-7
MDL Number: MFCD00012605
Molecular Formula: C9H13NO2
Molecular Weight: 203.67
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 143
Specific rotation [a]20/D: -45.5 deg (C=1, H2O)
Product Categories: Bioscience, Phenylethylamines
Other Product Names / Synonyms: (R)-(-)-3-[1-Hydroxy-2-(methylamino)ethyl]phenol Hydrochloride, (R)-(-)-1-(3-Hydroxyphenyl)-2-(methylamino)ethanol Hydrochloride, (R)-(-)-3-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol Hydrochloride
Formula:
C₉H₁₃NO₂·HCl |
MDL Number:
MFCD00012605 CAS Number: 61-76-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...