DL-Phenylephrine Hydrochloride ≥98.0% (by HPLC, titration analysis)

Synonyms: 3-[1-Hydroxy-2-(methylamino)ethyl]phenol Hydrochloride1-(3-Hydroxyphenyl)-2-(methylamino)ethanol Hydrochloride3-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol Hydrochloride

P0397-25G
TCP0397-025GEA 172.01 USD
TCP0397-025G
DL-Phenylephrine Hydrochloride ≥98.0% (by HPLC, titration analysis)
DL-Phenylephrine hydrochloride

CAS Number: 154-86-9
MDL Number: MFCD00070603
Molecular Formula: C9H13NO2
Molecular Weight: 203.67
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 144


Product Categories: Bioscience, Phenylethylamines
Other Product Names / Synonyms: 3-[1-Hydroxy-2-(methylamino)ethyl]phenol Hydrochloride, 1-(3-Hydroxyphenyl)-2-(methylamino)ethanol Hydrochloride, 3-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol Hydrochloride

Formula: C₉H₁₃NO₂
MW: 203.67 g/mol
MDL Number: MFCD00070603
CAS Number: 154-86-9

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR