Trioctyl phosphite (mixture) ≥85.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Phosphorous Acid Trioctyl Ester
P0386-25ML
TCP0386-025MLEA
49.73
USD
TCP0386-025ML
Trioctyl phosphite (mixture) ≥85.0% (by titrimetric analysis)
Trioctyl phosphite (mixture)
CAS Number: 3028-88-4
MDL Number: MFCD00059943
Molecular Formula: C24H51O3P
Molecular Weight: 418.64
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 182
Specific Gravity (20/20): 0.89
Other Product Names / Synonyms: Phosphorous Acid Trioctyl Ester
Formula:
C₂₄H₅₁O₃P MW: 418.64 g/mol Density: 0.89 (20 °C) Flash Pt: 182 °C |
MDL Number:
MFCD00059943 CAS Number: 3028-88-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...