Poly(1-vinylpyrrolidone-co-vinyl acetate) (copolymer 3:7) 50% in ethanol
Supplier: TCI America
P0385-500G
P0385-25G
TCP0385-500GEA
104.18
USD
TCP0385-500G
TCP0385-025G
Poly(1-vinylpyrrolidone-co-vinyl acetate) (copolymer 3:7) 50% in ethanol
Poly(1-vinylpyrrolidone-co-vinyl acetate)
CAS Number: 25086-89-9
MDL Number: MFCD00134018
Form: Clear Liquid
Color: Very Pale Yellow
Flash Point (°C): 13
Specific Gravity (20/20): 1.27
Product Categories: Materials Chemistry, Polymers
Density:
1.27 (20 °C) Flash Pt: 13 °C |
MDL Number:
MFCD00134018 CAS Number: 25086-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...