1-Phenyl-1-nonyne ≥98.0%
Supplier: TCI America
Synonyms:
1-Heptyl-2-phenylacetylene
P0363-5ML
TCP0363-005MLEA
264
USD
TCP0363-005ML
1-Phenyl-1-nonyne ≥98.0%
1-Phenyl-1-nonyne
CAS Number: 57718-18-0
MDL Number: MFCD00048944
Molecular Formula: C15H20
Molecular Weight: 200.33
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 0.89
Other Product Names / Synonyms: 1-Heptyl-2-phenylacetylene
Formula:
C₁₅H₂₀ MW: 200.33 g/mol Density: 0.89 (20 °C) |
MDL Number:
MFCD00048944 CAS Number: 57718-18-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...