Dimethyl phenylphosphonate ≥98.0%

Supplier: TCI America

Synonyms: Phenylphosphonic Acid Dimethyl EsterDimethyl BenzenephosphonateBenzenephosphonic Acid Dimethyl Ester

P0332-25ML
TCP0332-025MLEA 64.82 USD
TCP0332-025ML
Dimethyl phenylphosphonate ≥98.0%
Dimethyl phenylphosphonate

CAS Number: 2240-41-7
MDL Number: MFCD00059292
Molecular Formula: C8H11O3P
Molecular Weight: 186.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 247
Flash Point (°C): 154
Specific Gravity (20/20): 1.19


Other Product Names / Synonyms: Benzenephosphonic Acid Dimethyl Ester, Dimethyl Benzenephosphonate, Phenylphosphonic Acid Dimethyl Ester

Formula: C₈H₁₁O₃P
MW: 186.15 g/mol
Boiling Pt: 247 °C
Density: 1.19 (20 °C)
Flash Pt: 154 °C
MDL Number: MFCD00059292
CAS Number: 2240-41-7
https://us.vwr.com/stibo/web/std.lang.all/10/64/28991064.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR