(±)-1,2-Epithiopropane ≥98.0% stabilized

Supplier: TCI America

Synonyms: (±)-Propylene sulfide(±)-Propylene sulphide(±)-Propene sulfide(±)-Propene sulphideMethylethylene sulphidePropylene sulfidePropylene sulphideMethylethylene sulfide

P0321-25ML
TCP0321-025MLEA 220.3 USD
TCP0321-025ML
(±)-1,2-Epithiopropane ≥98.0% stabilized
(±)-1,2-Epithiopropane

(stabilized with Butyl Mercaptan)
CAS Number: 1072-43-1
MDL Number: MFCD00005160
Molecular Formula: C3H6S
Molecular Weight: 74.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 75
Flash Point (°C): 10
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Methylethylene Sulfide

Formula: C₃H₆S
MW: 74.15 g/mol
Boiling Pt: 75 °C
Melting Pt: –91 °C
Density: 0.94 (20 °C)
Flash Pt: 10 °C
MDL Number: MFCD00005160
CAS Number: 1072-43-1
EINECS: 214-007-3
https://us.vwr.com/stibo/web/std.lang.all/10/55/28991055.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR