Pentachlorothiophenol ≥95.0% (by titrimetric analysis)

Supplier: TCI America

P0312-25G
TCP0312-025GEA 282.09 USD
TCP0312-025G
Pentachlorothiophenol ≥95.0% (by titrimetric analysis)
Pentachlorothiophenol

CAS Number: 133-49-3
MDL Number: MFCD00059146
Molecular Formula: C6HCl5S
Molecular Weight: 282.38
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 242


Other Product Names / Synonyms: Pentachlorothiophenol

Formula: C₆HCl₅S
Melting Pt: 242 °C
MDL Number: MFCD00059146
CAS Number: 133-49-3
https://us.vwr.com/stibo/web/std.lang.all/10/41/28991041.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR