6-Phenyl-2-thiouracil ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Mercapto-6-phenylpyrimidin-4-ol

P0236-5G P0236-1G
TCP0236-001GEA 149 USD
TCP0236-001G TCP0236-005G
6-Phenyl-2-thiouracil ≥98.0% (by HPLC, titration analysis)
6-Phenyl-2-thiouracil

CAS Number: 36822-11-4
MDL Number: MFCD00059166
Molecular Formula: C10H8N2OS
Molecular Weight: 204.25
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

Formula: C₁₀H₈N₂OS
MW: 204.25 g/mol
MDL Number: MFCD00059166
CAS Number: 36822-11-4
https://us.vwr.com/stibo/web/std.lang.all/09/66/28990966.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR