N-Phenylthiobenzamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

P0233-25G P0233-5G
TCP0233-005GEA 182 USD
TCP0233-005G TCP0233-025G
N-Phenylthiobenzamide ≥98.0% (by HPLC, total nitrogen)
N-Phenylthiobenzamide

CAS Number: 636-04-4
MDL Number: MFCD00022147
Molecular Formula: C13H11NS
Molecular Weight: 213.30
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 102


Other Product Names / Synonyms: Thiobenzanilide

Formula: C₁₃H₁₁NS
MW: 213.2981 g/mol
Melting Pt: 102 °C
MDL Number: MFCD00022147
CAS Number: 636-04-4
https://us.vwr.com/stibo/web/std.lang.all/09/64/28990964.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR