1,2-Phenylenediamine sulfate
Supplier: TCI America
Synonyms:
1,2-Diaminobenzene Sulphate
, 1,2-Diaminobenzene Sulfate
P0173-25G
TCP0173-025GEA
112.62
USD
TCP0173-025G
1,2-Phenylenediamine sulfate
1,2-Phenylenediamine sulfate
CAS Number: 74710-09-1
MDL Number: MFCD00054335
Molecular Formula: C6H8N2
Molecular Weight: 206.22
Form: Crystal
Color: White
Other Product Names / Synonyms: 1,2-Diaminobenzene Sulfate
Formula:
C₆H₈N₂ MW: 206.22 g/mol |
MDL Number:
MFCD00054335 CAS Number: 74710-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...