2-Phenylazo-4-methylphenol ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
2-Phenylazo-p-cresol
, 4-Methyl-2-phenylazophenol
, 2-Hydroxy-5-methylazobenzene
P0145-25G
TCP0145-025GEA
597
USD
TCP0145-025G
2-Phenylazo-4-methylphenol ≥98.0% (by HPLC, total nitrogen)
2-Phenylazo-4-methylphenol
CAS Number: 952-47-6
MDL Number: MFCD00045788
Molecular Formula: C13H12N2O
Molecular Weight: 212.25
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 105
Other Product Names / Synonyms: 2-Hydroxy-5-methylazobenzene, 4-Methyl-2-phenylazophenol, 2-Phenylazo-p-cresol
Formula:
C₁₃H₁₂N₂O MW: 212.25 g/mol Melting Pt: 105 °C |
MDL Number:
MFCD00045788 CAS Number: 952-47-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...