4-Hydroxybenzenesulfonic acid ≥85.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: p-Phenolsulfonic acidPhenol-4-sulfonic acid4-Hydroxybenzenesulfonic acid hydratePhenol-4-sulphonic acid4-Hydroxybenzenesulphonic acid hydratep-Phenolsulphonic acid

P0104-25G P0104-500G
TCP0104-025GEA 23.35 USD
TCP0104-025G TCP0104-500G
4-Hydroxybenzenesulfonic acid ≥85.0% (by titrimetric analysis)
4-Hydroxybenzenesulphonic acid

CAS Number: 98-67-9
MDL Number: MFCD00673723
Molecular Formula: C6H6O4S
Molecular Weight: 174.17
Purity/Analysis Method: 85.0% (T)
Form: Crystal


Other Product Names / Synonyms: p-Phenolsulfonic Acid, Phenol-4-sulfonic Acid

Formula: C₆H₆O₄S
MW: 174.17
Storage Temperature: Ambient
MDL Number: MFCD00673723
CAS Number: 98-67-9
EINECS: 202-691-6
https://us.vwr.com/stibo/web/std.lang.all/07/98/28990798.jpg

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR