n-Pentatriacontane ≥95.0%

Supplier: TCI America

P0062-1G P0062-10G
TCP0062-010GEA 1195.32 USD
TCP0062-010G TCP0062-001G
n-Pentatriacontane ≥95.0%
n-Pentatriacontane

CAS Number: 630-07-9
MDL Number: MFCD00059397
Molecular Formula: C35H72
Molecular Weight: 492.96
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Boiling point (°C): 331
Melting point (°C): 74

Formula: H₃C(CH₂)₃₃CH₃
MW: 492.96 g/mol
Boiling Pt: 331 °C
Melting Pt: 74 °C
Density: 0.8157 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00059397
CAS Number: 630-07-9
https://us.vwr.com/stibo/web/std.lang.all/07/69/28990769.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR