Palmitic anhydride ≥96.0% (by GC, titration analysis)

Supplier: TCI America

P0008-500G P0008-25G
TCP0008-500GEA 447 USD
TCP0008-500G TCP0008-025G
Palmitic anhydride ≥96.0% (by GC, titration analysis)
Palmitic anhydride

CAS Number: 623-65-4
MDL Number: MFCD00008992
Molecular Formula: C32H62O3
Molecular Weight: 494.85
Purity/Analysis Method: 96.0% (GC,T)
Form: Crystal
Melting point (°C): 64

https://us.vwr.com/stibo/web/std.lang.all/07/35/28990735.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR