Tributyloctylphosphonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tributyl(octyl)phosphonium bromide

O0297-25G
TCO0297-25GEA 152 USD
TCO0297-25G
Tributyloctylphosphonium bromide ≥98.0% (by titrimetric analysis)
Tributyloctylphosphonium bromide

CAS Number: 57702-65-5
MDL Number: MFCD04038147
Molecular Formula: C20H44BrP
Molecular Weight: 395.45
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Freezing point (°C): 25


Other Product Names / Synonyms: n-Octyltributylphosphonium Bromide

Formula: C₂₀H₄₄BrP
MW: 395.4411 g/mol
MDL Number: MFCD04038147
CAS Number: 57702-65-5
https://us.vwr.com/stibo/web/std.lang.all/05/69/28990569.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR