Butamben (butyl-4-aminobenzoate) ≥99.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Butyl 4-aminobenzoate
A0270-25G
TCA0270-025GEA
24.36
USD
TCA0270-025G
Butamben (butyl-4-aminobenzoate) ≥99.0% (by GC, titration analysis)
Butamben (Butyl 4-aminobenzoate)
CAS Number: 94-25-7
MDL Number: MFCD00017112
Molecular Formula: C11H15NO2
Molecular Weight: 193.25
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 174
Melting point (°C): 60
Other Product Names / Synonyms: 4-Aminobenzoic Acid Butyl Ester
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...