4-Amino-3-bromobenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A1969-5G
TCA1969-5GEA
84.76
USD
TCA1969-5G
4-Amino-3-bromobenzoic acid ≥98.0% (by titrimetric analysis)
4-Amino-3-bromobenzoic acid
CAS Number: 6311-37-1
MDL Number: MFCD03407439
Molecular Formula: C7H6BrNO2
Molecular Weight: 216.03
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 212
Formula:
C₇H₆BrNO₂ MW: 216.0320 g/mol Melting Pt: 212 °C |
MDL Number:
MFCD03407439 CAS Number: 6311-37-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...