Dichloro(methyl)octadecylsilane ≥94.0%

Supplier: TCI America

Synonyms: Dichlorooctadecylmethylsilane

O0264-25ML
TCO0264-025MLEA 100.45 USD
TCO0264-025ML
Dichloro(methyl)octadecylsilane ≥94.0%
Dichloro(methyl)octadecylsilane

CAS Number: 5157-75-5
MDL Number: MFCD00018083
Molecular Formula: C19H40Cl2Si
Molecular Weight: 367.51
Purity/Analysis Method: 94.0% (GC)
Form: Crystal
Boiling point (°C): 210
Flash Point (°C): 185
Freezing point (°C): 23


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: Dichlorooctadecylmethylsilane

Formula: C₁₉H₄₀Cl₂Si
MW: 367.51 g/mol
Boiling Pt: 210 °C
Flash Pt: 185 °C
MDL Number: MFCD00018083
CAS Number: 5157-75-5
https://us.vwr.com/stibo/web/std.lang.all/05/35/28990535.jpg

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR