3-Octadecylthiophene ≥98.0%

Supplier: TCI America

O0245-5G O0245-1G
TCO0245-001GEA 46 USD
TCO0245-001G TCO0245-005G
3-Octadecylthiophene ≥98.0%
3-Octadecylthiophene

CAS Number: 104934-54-5
MDL Number: MFCD00191567
Molecular Formula: C22H40S
Molecular Weight: 336.62
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 36
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Building Blocks for Semiconducting Polymers, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-Stearylthiophene

Formula: C₂₂H₄₀S
Melting Pt: 36 °C
MDL Number: MFCD00191567
CAS Number: 104934-54-5
https://us.vwr.com/stibo/web/std.lang.all/05/21/28990521.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR