(2,7-Octadien-1-yl)succinic anhydride
Supplier: TCI America
Synonyms:
4,9-Decadiene-1,2-dicarboxylic Anhydride
O0244-25G
TCO0244-025GEA
389.64
USD
TCO0244-025G
(2,7-Octadien-1-yl)succinic anhydride
(2,7-Octadien-1-yl)succinic anhydride
CAS Number: 142826-45-7
MDL Number: MFCD00191676
Molecular Formula: C12H16O3
Molecular Weight: 208.26
Form: Clear Liquid
Color: Very Pale Yellow
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: 4,9-Decadiene-1,2-dicarboxylic Anhydride
Formula:
C₁₂H₁₆O₃ MW: 208.26 g/mol Density: 1.05 (20 °C) |
MDL Number:
MFCD00191676 CAS Number: 142826-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...